missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals

29.75 € - 722.60 €

Chemical Identifiers

CAS 121-88-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00007692
InChI Key DOPJTDJKZNWLRB-UHFFFAOYSA-N
Synonym 2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene
PubChem CID 4984721
ChEBI CHEBI:82384
IUPAC Name 2-amino-5-nitrophenol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)N
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10552294
View Documents
Thermo Scientific Acros
400900250
25 g Glass bottle
29.75 €
25g
Please to purchase this item. Need a web account? Register with us today!
10340664
View Documents
Thermo Scientific Acros
400900500
50 g Glass bottle
48.94 €
50g
Please to purchase this item. Need a web account? Register with us today!
10229740
View Documents
Thermo Scientific Acros
400902500
250 g Glass bottle
198.99 €
250g
Please to purchase this item. Need a web account? Register with us today!
10455085
View Documents
Thermo Scientific Acros
400900010
1 kg Glass bottle
722.60 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-88-0
154.13
DOPJTDJKZNWLRB-UHFFFAOYSA-N
4984721
2-amino-5-nitrophenol
C6H6N2O3
MFCD00007692
2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene
CHEBI:82384
C1=CC(=C(C=C1[N+](=O)[O-])O)N

Specifications

121-88-0
100.0
Brown to Brown-Rust
95%
MFCD00007692
Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene and diethylether
C1=CC(=C(C=C1[N+](=O)[O-])O)N
154.13
CHEBI:82384
95%
2-Amino-5-nitrophenol, 95%
94.0
198.0°C to 202.0°C
Authentic
C6H6N2O3
2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene
DOPJTDJKZNWLRB-UHFFFAOYSA-N
2-amino-5-nitrophenol
4984721
154.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 204-503-8

missing translation for 'rtecsNumber' : SJ6302500

missing translation for 'tsca' : TSCA

Product Identifier
  • 2-Amino-5-nitrophenol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.