missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorobenzothiazole, 97%, Thermo Scientific™

Product Code. 11463677
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19952-47-7
Molecular Formula C7H5ClN2S
Molecular Weight (g/mol) 184.641
MDL Number MFCD00005791
InChI Key OEQQFQXMCPMEIH-UHFFFAOYSA-N
Synonym 2-amino-4-chlorobenzothiazole, 2-benzothiazolamine, 4-chloro, 4-chlorobenzo d thiazol-2-amine, 4-chlorobenzothiazol-2-ylamine, benzothiazole, 2-amino-4-chloro, 4-chloro-2-benzothiazolamine, 4-chloro-2-aminobenzothiazole, 4-chloro-benzothiazol-2-ylamine, 4-chlorobenzothiazole-2-ylamine, zlchem 166
PubChem CID 29872
IUPAC Name 4-chloro-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C(=C1)Cl)N=C(S2)N

Specifications

Melting Point 203°C to 205°C
Quantity 25 g
Beilstein 136784
Formula Weight 184.65
Percent Purity 97%
Chemical Name or Material 2-Amino-4-chlorobenzothiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'