Learn More
2-Amino-4,6-dimethylpyrimidine, 98%, Thermo Scientific Chemicals
28.94 € - 85.63 €
Chemical Identifiers
CAS | 767-15-7 |
---|---|
Molecular Formula | C6H9N3 |
Molecular Weight (g/mol) | 123.16 |
MDL Number | MFCD00006102 |
InChI Key | IDQNBVFPZMCDDN-UHFFFAOYSA-N |
Synonym | 2-amino-4,6-dimethylpyrimidine, acetylacetoneguanidine, 2-pyrimidinamine, 4,6-dimethyl, 4,6-dimethyl-2-pyrimidinamine, 4,6-dimethylpyrimidin-2-ylamine, 4,6-dimethyl-2-aminopyrimidine, pyrimidine, 2-amino-4,6-dimethyl, unii-4s48mi253m, 2-amine-4,6-dimethylpyrimidine, 2-amino-4,6-dimethyl pyrimidine |
PubChem CID | 13021 |
IUPAC Name | 4,6-dimethylpyrimidin-2-amine |
SMILES | CC1=CC(C)=NC(N)=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11420284
|
Thermo Scientific Alfa Aesar
A13674.14 |
25 g |
28.94 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430284
|
Thermo Scientific Alfa Aesar
A13674.22 |
100 g |
85.63 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
767-15-7 | |
123.16 | |
IDQNBVFPZMCDDN-UHFFFAOYSA-N | |
13021 | |
CC1=CC(C)=NC(N)=N1 |
C6H9N3 | |
MFCD00006102 | |
2-amino-4,6-dimethylpyrimidine, acetylacetoneguanidine, 2-pyrimidinamine, 4,6-dimethyl, 4,6-dimethyl-2-pyrimidinamine, 4,6-dimethylpyrimidin-2-ylamine, 4,6-dimethyl-2-aminopyrimidine, pyrimidine, 2-amino-4,6-dimethyl, unii-4s48mi253m, 2-amine-4,6-dimethylpyrimidine, 2-amino-4,6-dimethyl pyrimidine | |
4,6-dimethylpyrimidin-2-amine |
Specifications
767-15-7 | |
C6H9N3 | |
110735 | |
IDQNBVFPZMCDDN-UHFFFAOYSA-N | |
4,6-dimethylpyrimidin-2-amine | |
13021 | |
98% |
150°C to 155°C | |
MFCD00006102 | |
2-amino-4,6-dimethylpyrimidine, acetylacetoneguanidine, 2-pyrimidinamine, 4,6-dimethyl, 4,6-dimethyl-2-pyrimidinamine, 4,6-dimethylpyrimidin-2-ylamine, 4,6-dimethyl-2-aminopyrimidine, pyrimidine, 2-amino-4,6-dimethyl, unii-4s48mi253m, 2-amine-4,6-dimethylpyrimidine, 2-amino-4,6-dimethyl pyrimidine | |
CC1=CC(C)=NC(N)=N1 | |
123.16 | |
123.16 | |
2-Amino-4,6-dimethylpyrimidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 212-181-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 2-Amino-4,6-dimethylpyrimidine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 2-Amino-4,6-dimethylpyrimidine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.