Learn More
2'-Amino-3'-methoxyflavone, 99%, Thermo Scientific Chemicals
A MAP kinase inhibitor
88.65 € - 310.00 €
Chemical Identifiers
CAS | 167869-21-8 |
---|---|
Molecular Formula | C16H13NO3 |
Molecular Weight (g/mol) | 267.28 |
MDL Number | MFCD00671789 |
InChI Key | QFWCYNPOPKQOKV-UHFFFAOYSA-N |
Synonym | 2-2-amino-3-methoxyphenyl-4h-chromen-4-one, 2-2-amino-3-methoxyphenyl-4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl chromen-4-one, 2'-amino-3'-methoxyflavone, unii-sje1io5e3i, sje1io5e3i, chembl35482, 2-2-amino-3-methoxyphenyl-chromen-4-one, 4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl, 2′-amino-3′-methoxyflavone |
PubChem CID | 4713 |
ChEBI | CHEBI:77954 |
IUPAC Name | 2-(2-amino-3-methoxyphenyl)chromen-4-one |
SMILES | COC1=CC=CC(=C1N)C1=CC(=O)C2=CC=CC=C2O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15446419
|
Thermo Scientific Alfa Aesar
J64485.#0 |
5 mg | |||||||
15456419
|
Thermo Scientific Alfa Aesar
J64485.MA |
10 mg |
148.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15466419
|
Thermo Scientific Alfa Aesar
J64485.MB |
25 mg |
310.00 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A MAP kinase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
167869-21-8 | |
267.28 | |
QFWCYNPOPKQOKV-UHFFFAOYSA-N | |
4713 | |
2-(2-amino-3-methoxyphenyl)chromen-4-one |
C16H13NO3 | |
MFCD00671789 | |
2-2-amino-3-methoxyphenyl-4h-chromen-4-one, 2-2-amino-3-methoxyphenyl-4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl chromen-4-one, 2'-amino-3'-methoxyflavone, unii-sje1io5e3i, sje1io5e3i, chembl35482, 2-2-amino-3-methoxyphenyl-chromen-4-one, 4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl, 2′-amino-3′-methoxyflavone | |
CHEBI:77954 | |
COC1=CC=CC(=C1N)C1=CC(=O)C2=CC=CC=C2O1 |
Specifications
167869-21-8 | |
C16H13NO3 | |
2-2-amino-3-methoxyphenyl-4h-chromen-4-one, 2-2-amino-3-methoxyphenyl-4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl chromen-4-one, 2'-amino-3'-methoxyflavone, unii-sje1io5e3i, sje1io5e3i, chembl35482, 2-2-amino-3-methoxyphenyl-chromen-4-one, 4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl, 2′-amino-3′-methoxyflavone | |
COC1=CC=CC(=C1N)C1=CC(=O)C2=CC=CC=C2O1 | |
267.28 | |
CHEBI:77954 | |
99% | |
2'-Amino-3'-methoxyflavone |
Yellow | |
MFCD00671789 | |
QFWCYNPOPKQOKV-UHFFFAOYSA-N | |
2-(2-amino-3-methoxyphenyl)chromen-4-one | |
4713 | |
267.28 | |
Powder |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
Recommended Storage : −30° to −10°C
RUO – Research Use Only