missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (2-Allylphenoxy)trimethylsilane, 98%, Thermo Scientific™

Product Code. 15493997
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18042-43-8
Molecular Formula C12H18OSi
Molecular Weight (g/mol) 206.36
MDL Number MFCD12407078
InChI Key LJIXCUHZKDSCQX-UHFFFAOYSA-N
Synonym 2-allylphenoxy trimethylsilane, trimethyl 2-prop-2-en-1-yl phenoxy silane, o-allylphenoxytrimethylsilane, trimethyl-2-prop-2-enylphenoxy silane, silane, trimethyl 2-2-propenyl phenoxy
PubChem CID 15049460
IUPAC Name trimethyl-(2-prop-2-enylphenoxy)silane
SMILES C[Si](C)(C)OC1=CC=CC=C1CC=C

Specifications

Boiling Point 85°C (5mmHg)
Quantity 25 g
Formula Weight 206.36
Percent Purity 98%
Sensitivity Moisture sensitive
Density 0.925
Chemical Name or Material (2-Allylphenoxy)trimethylsilane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'