missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanamine hydrochloride, 98+%, Thermo Scientific Chemicals

25.70 € - 260.00 €

Chemical Identifiers

CAS 10523-68-9
Molecular Formula C10H18ClN
Molecular Weight (g/mol) 187.71
MDL Number MFCD00074743
InChI Key WLDWDRZITJEWRJ-UHFFFAOYSA-N
Synonym 2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride
PubChem CID 25331
SMILES [H+].[Cl-].NC1C2CC3CC(C2)CC1C3
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11464040
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17140.03
1 g
25.70 €
1g
Please to purchase this item. Need a web account? Register with us today!
11483357
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17140.06
5 g
81.80 €
5g
Please to purchase this item. Need a web account? Register with us today!
11493357
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A17140.14
25 g
260.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Adamantylamine hydrochloride was used to prepare 2-adamantylamide of 2?-(carboxymethoxime)-olivomycin I. The HPLC assay of 2-adamantylamine hydrochloride after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole has been studied. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Adamantylamine hydrochloride was used to prepare 2-adamantylamide of 2′-(carboxymethoxime)-olivomycin I. The HPLC assay of 2-adamantylamine hydrochloride after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole has been studied. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

10523-68-9
187.71
WLDWDRZITJEWRJ-UHFFFAOYSA-N
25331
C10H18ClN
MFCD00074743
2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride
[H+].[Cl-].NC1C2CC3CC(C2)CC1C3

Specifications

10523-68-9
C10H18ClN
4297901
Soluble in water.
[H+].[Cl-].NC1C2CC3CC(C2)CC1C3
187.71
187.71
2-Adamantanamine hydrochloride
>300°C
MFCD00074743
2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride
WLDWDRZITJEWRJ-UHFFFAOYSA-N
adamantan-2-amine;hydrochloride
25331
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 234-074-2

missing translation for 'rtecsNumber' : AU4375100

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only