missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylpyrrole, 98%, Thermo Scientific Chemicals

Product Code. 11402018
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-83-9
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00005220
InChI Key IGJQUJNPMOYEJY-UHFFFAOYSA-N
Synonym 2-acetylpyrrole, 1-1h-pyrrol-2-yl ethanone, 1-1h-pyrrol-2-yl ethan-1-one, 2-acetyl pyrrole, 2-pyrrolylethanone, 2-acetyl-1h-pyrrole, methyl pyrrol-2-yl ketone, ethanone, 1-1h-pyrrol-2-yl, 2-pyrrolyl methyl ketone, ketone, methyl pyrrol-2-yl
PubChem CID 14079
ChEBI CHEBI:59981
IUPAC Name 1-(1H-pyrrol-2-yl)ethanone
SMILES CC(=O)C1=CC=CN1

Specifications

Melting Point 88°C to 92°C
Boiling Point 220°C
Odor Characteristic
Quantity 5 g
Beilstein 1882
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2-Acetylpyrrole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'