Learn More
2-Acetylpyridine, 98%, Thermo Scientific Chemicals
46.33 € - 525.62 €
Chemical Identifiers
CAS | 1122-62-9 |
---|---|
Molecular Formula | C7H7NO |
Molecular Weight (g/mol) | 121.14 |
InChI Key | AJKVQEKCUACUMD-UHFFFAOYSA-N |
Synonym | 2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone |
PubChem CID | 14286 |
IUPAC Name | 1-pyridin-2-ylethanone |
SMILES | CC(=O)C1=CC=CC=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10408630
|
Thermo Scientific Acros
102680250 |
25 g | Glass bottle |
46.33 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10346260
|
Thermo Scientific Acros
102681000 |
100 g | Glass bottle |
134.67 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11966281
|
Thermo Scientific Acros
102685000 |
500 g | Glass bottle |
525.62 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1122-62-9 | |
121.14 | |
2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone | |
1-pyridin-2-ylethanone |
C7H7NO | |
AJKVQEKCUACUMD-UHFFFAOYSA-N | |
14286 | |
CC(=O)C1=CC=CC=N1 |
Specifications
1122-62-9 | |
100.0 | |
Colorless to Brown | |
188.0°C to 189.0°C | |
Authentic | |
C7H7NO | |
1.5200 to 1.5220 | |
2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone | |
AJKVQEKCUACUMD-UHFFFAOYSA-N | |
1-pyridin-2-ylethanone | |
14286 | |
98% | |
2-Acetylpyridine |
97.5 | |
8.0°C to 10.0°C | |
1.0800g/mL | |
76°C | |
97.5% min. (GC) | |
C5H4NCOCH3 | |
1.08 | |
Solubility in water: 200g/L (20°C). Other solubilities: miscible with ethanol | |
CC(=O)C1=CC=CC=N1 | |
121.14 | |
121.14 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 214-355-6
- 2-Acetylpyridine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.