missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbenzo[b]furan, 99%, Thermo Scientific Chemicals

Product Code. 11333237
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Acetylbenzo[b]furan is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylbenzo[b]furan is used as a fine chemical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1646-26-0
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00005849
InChI Key YUTFQTAITWWGFH-UHFFFAOYSA-N
Synonym 2-acetylbenzofuran, 1-benzofuran-2-yl ethanone, ethanone, 1-2-benzofuranyl, benzofuran-2-yl methyl ketone, 2-acetylcoumarone, 2-acetylcumarone, 2-benzofuranyl methyl ketone, benzo b furan-2-yl methyl ketone, 1-2-benzofuranyl ethanone, 2-acetylbenzo b furan
PubChem CID 15435
SMILES CC(=O)C1=CC2=CC=CC=C2O1

Specifications

Melting Point 70°C to 72°C
Boiling Point 110°C to 113°C (3 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 117910
Solubility Information Insoluble in water.
IUPAC Name 1-(1-benzofuran-2-yl)ethanone
Formula Weight 160.17
Percent Purity 99%
Chemical Name or Material 2-Acetylbenzo[b]furan

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'