Learn More
2-Acetyl-5-methylfuran, 98+%, Thermo Scientific Chemicals
39.90 € - 114.00 €
Chemical Identifiers
CAS | 1193-79-9 |
---|---|
Molecular Formula | C7H8O2 |
Molecular Weight (g/mol) | 124.139 |
MDL Number | MFCD00003243 |
InChI Key | KEFJLCGVTHRGAH-UHFFFAOYSA-N |
Synonym | 2-acetyl-5-methylfuran, 5-methyl-2-acetylfuran, 1-5-methylfuran-2-yl ethanone, 1-5-methyl-2-furyl ethanone, 5-methyl-2-furylmethylketone, ethanone, 1-5-methyl-2-furanyl, 1-5-methyl-2-furanyl ethanone, 2-methyl-5-acetylfuran, 1-5-methyl-2-furyl ethan-1-one, methyl 5-methyl-2-furyl ketone |
PubChem CID | 14514 |
ChEBI | CHEBI:562752 |
IUPAC Name | 1-(5-methylfuran-2-yl)ethanone |
SMILES | CC1=CC=C(O1)C(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11423237
|
Thermo Scientific Alfa Aesar
A16172.09 |
10 g |
39.90 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494040
|
Thermo Scientific Alfa Aesar
A16172.14 |
25 g |
68.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433237
|
Thermo Scientific Alfa Aesar
A16172.18 |
50 g |
114.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlucose was transformed to various furan compounds and carboxylic acids derivatives by heat treatment of autoclave such as levulinic acid, 2-acetylfuran and 2-acetyl-5-methylfuran. using 2-acetyl-5-methylfuran or 1,5-dimethyl-2-pyrrolecarbonitrile as reactants, that the formation of the 4-arylation products. Reaction of 2-acetyl-5-methylfuran 6a with 2-aminoethanol pioduced in 79% yield the aminal.
Solubility
Slightly soluble in water. Soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
1193-79-9 | |
124.139 | |
KEFJLCGVTHRGAH-UHFFFAOYSA-N | |
14514 | |
1-(5-methylfuran-2-yl)ethanone |
C7H8O2 | |
MFCD00003243 | |
2-acetyl-5-methylfuran, 5-methyl-2-acetylfuran, 1-5-methylfuran-2-yl ethanone, 1-5-methyl-2-furyl ethanone, 5-methyl-2-furylmethylketone, ethanone, 1-5-methyl-2-furanyl, 1-5-methyl-2-furanyl ethanone, 2-methyl-5-acetylfuran, 1-5-methyl-2-furyl ethan-1-one, methyl 5-methyl-2-furyl ketone | |
CHEBI:562752 | |
CC1=CC=C(O1)C(=O)C |
Specifications
1193-79-9 | |
100°C to 101°C (25 mmHg) | |
C7H8O2 | |
MFCD00003243 | |
2-acetyl-5-methylfuran, 5-methyl-2-acetylfuran, 1-5-methylfuran-2-yl ethanone, 1-5-methyl-2-furyl ethanone, 5-methyl-2-furylmethylketone, ethanone, 1-5-methyl-2-furanyl, 1-5-methyl-2-furanyl ethanone, 2-methyl-5-acetylfuran, 1-5-methyl-2-furyl ethan-1-one, methyl 5-methyl-2-furyl ketone | |
KEFJLCGVTHRGAH-UHFFFAOYSA-N | |
1-(5-methylfuran-2-yl)ethanone | |
14514 | |
124.14 | |
2-Acetyl-5-methylfuran |
1.068 | |
80°C (176°F) | |
1.513 | |
110853 | |
Slightly soluble in water. Soluble in alcohol. | |
CC1=CC=C(O1)C(=O)C | |
124.139 | |
CHEBI:562752 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
H227-H302
missing translation for 'einecsNumber' : 214-779-1
missing translation for 'rtecsNumber' : LT8528000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.