missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-3-methylpyrazine, 98%, Thermo Scientific Chemicals

Product Code. 11477327
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23787-80-6
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00014612
InChI Key QUNOTZOHYZZWKQ-UHFFFAOYSA-N
Synonym 2-acetyl-3-methylpyrazine, 1-3-methylpyrazin-2-yl ethanone, 1-3-methylpyrazin-2-yl ethan-1-one, ethanone, 1-3-methylpyrazinyl, 2-methyl-3-acetylpyrazine, unii-7u5tew2y2b, 2-acetyl-3-methyl pyrazine, 1-3-methylpyrazinyl ethan-1-one, 3-acetyl-2-methylpyrazine, ethanone, 1-3-methyl-2-pyrazinyl
PubChem CID 32093
IUPAC Name 1-(3-methylpyrazin-2-yl)ethanone
SMILES CC(=O)C1=NC=CN=C1C

Specifications

Density 1.105
Boiling Point 265°C to 266°C
Flash Point >100°C (212°F)
Refractive Index 1.522
Quantity 5 g
Beilstein 742438
Sensitivity Light sensitive
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 2-Acetyl-3-methylpyrazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'