Learn More
2-Acetyl-3-bromothiophene, 97%, Thermo Scientific™
21.90 € - 120.79 €
Chemical Identifiers
CAS | 42877-08-7 |
---|---|
Molecular Formula | C6H5BrOS |
Molecular Weight (g/mol) | 205.07 |
MDL Number | MFCD00794287 |
InChI Key | ZUXPELAHJQSZTE-UHFFFAOYSA-N |
Synonym | 2-acetyl-3-bromothiophene, 1-3-bromothiophen-2-yl ethanone, 1-3-bromothiophen-2-yl ethan-1-one, 1-3-bromo-2-thienyl-1-ethanone, bromothienylethanone, 1-3-bromothien-2-yl ethanone, 1-3-bromo-2-thienyl ethanone, pubchem7280, acmc-20ai7w |
PubChem CID | 2764496 |
SMILES | CC(=O)C1=C(Br)C=CS1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11354556
|
Thermo Scientific Alfa Aesar
L20113.03 |
1 g |
21.90 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11364556
|
Thermo Scientific Alfa Aesar
L20113.06 |
5 g |
37.69 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11374556
|
Thermo Scientific Alfa Aesar
L20113.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
42877-08-7 | |
205.07 | |
ZUXPELAHJQSZTE-UHFFFAOYSA-N | |
2764496 |
C6H5BrOS | |
MFCD00794287 | |
2-acetyl-3-bromothiophene, 1-3-bromothiophen-2-yl ethanone, 1-3-bromothiophen-2-yl ethan-1-one, 1-3-bromo-2-thienyl-1-ethanone, bromothienylethanone, 1-3-bromothien-2-yl ethanone, 1-3-bromo-2-thienyl ethanone, pubchem7280, acmc-20ai7w | |
CC(=O)C1=C(Br)C=CS1 |
Specifications
42877-08-7 | |
102°C to 105°C (4 mmHg) | |
1.614 | |
116973 | |
2-acetyl-3-bromothiophene, 1-3-bromothiophen-2-yl ethanone, 1-3-bromothiophen-2-yl ethan-1-one, 1-3-bromo-2-thienyl-1-ethanone, bromothienylethanone, 1-3-bromothien-2-yl ethanone, 1-3-bromo-2-thienyl ethanone, pubchem7280, acmc-20ai7w | |
CC(=O)C1=C(Br)C=CS1 | |
205.07 | |
205.07 | |
2-Acetyl-3-bromothiophene |
41°C to 45°C | |
C6H5BrOS | |
MFCD00794287 | |
Light sensitive | |
ZUXPELAHJQSZTE-UHFFFAOYSA-N | |
1-(3-bromothiophen-2-yl)ethanone | |
2764496 | |
97% |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.