missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Acetamido-2-deoxy-D-glucopyranose, 98%

Product Code. 10063863
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00061615
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
Synonym N-Acetyl-D-glucosamine
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Loss on Drying 2% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 500 g
Merck Index 15, 4494
Specific Rotation Condition + 42.00 (20.00°C c=2,water,2 hrs)
Specific Rotation +39° to +45° (20°C, 589nm) (c=2, H2O, after 2 hrs)
Formula Weight 221.21
Percent Purity ≥97.5% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-2-deoxy-D-glucopyranose
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'