missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedicarboxylic acid chloride, 97%, Thermo Scientific Chemicals

Product Code. 10249643
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3739-94-4
Molecular Formula C7H3Cl2NO2
Molecular Weight (g/mol) 204.01
MDL Number MFCD00006289
InChI Key GWHOGODUVLQCEB-UHFFFAOYSA-N
Synonym 2,6-pyridinedicarbonyl dichloride, 2,6-pyridinedicarboxylic acid chloride, pyridine-2,6-dicarbonyl dichloride, 2,6-pyridinedicarbonyl chloride, pyridine-2,6-dicarboxylic acid chloride, 2,6-pyridinedicarboxylicacidchloride, pyridine-2,6-dicarboyl dichloride, 2,6-pyridinedicarbony chloride, pubchem13243, tpc-i153
PubChem CID 77335
IUPAC Name pyridine-2,6-dicarbonyl chloride
SMILES C1=CC(=NC(=C1)C(=O)Cl)C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 57.0°C to 61.0°C
Color White to Brown
Boiling Point 284.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C5H3N)(COCl)2
Quantity 100 g
Beilstein 22, 155
Formula Weight 204.01
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2, 6-Pyridinedicarboxylic acid chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Pyridinedicarboxylic acid chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.