Learn More
2,6-Dimethylmorpholine, cis + trans, 97%, Thermo Scientific Chemicals
55.60 € - 195.00 €
Chemical Identifiers
CAS | 141-91-3 |
---|---|
Molecular Formula | C6H13NO |
Molecular Weight (g/mol) | 115.176 |
MDL Number | MFCD00005973 |
InChI Key | HNVIQLPOGUDBSU-UHFFFAOYSA-N |
Synonym | morpholine, 2,6-dimethyl, 2,6-dimethyl morpholine, 2,6-dimethylmorfolin, 2,6-dimethylmorfolin czech, ccris 5910, trans-2,6-dimethylmorpholine, 2,6-dimethyl-2,3,5,6-tetrahydro-4h-1,4-oxazine, 2,6-dimethyl-morpholine, 2,6-dimethylmorpholine, cis + trans, acmc-209nod |
PubChem CID | 110862 |
IUPAC Name | 2,6-dimethylmorpholine |
SMILES | CC1CNCC(O1)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435730
|
Thermo Scientific Alfa Aesar
H25954.14 |
25 g | |||||||
11445730
|
Thermo Scientific Alfa Aesar
H25954.22 |
100 g |
195.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-91-3 | |
115.176 | |
HNVIQLPOGUDBSU-UHFFFAOYSA-N | |
110862 | |
CC1CNCC(O1)C |
C6H13NO | |
MFCD00005973 | |
morpholine, 2,6-dimethyl, 2,6-dimethyl morpholine, 2,6-dimethylmorfolin, 2,6-dimethylmorfolin czech, ccris 5910, trans-2,6-dimethylmorpholine, 2,6-dimethyl-2,3,5,6-tetrahydro-4h-1,4-oxazine, 2,6-dimethyl-morpholine, 2,6-dimethylmorpholine, cis + trans, acmc-209nod | |
2,6-dimethylmorpholine |
Specifications
141-91-3 | |
0.935 | |
48°C (120°F) | |
1.446 | |
UN1992 | |
morpholine, 2,6-dimethyl, 2,6-dimethyl morpholine, 2,6-dimethylmorfolin, 2,6-dimethylmorfolin czech, ccris 5910, trans-2,6-dimethylmorpholine, 2,6-dimethyl-2,3,5,6-tetrahydro-4h-1,4-oxazine, 2,6-dimethyl-morpholine, 2,6-dimethylmorpholine, cis + trans, acmc-209nod | |
CC1CNCC(O1)C | |
115.176 | |
115.18 | |
2,6-Dimethylmorpholine, cis + trans |
-85°C | |
147°C | |
C6H13NO | |
MFCD00005973 | |
103036 | |
HNVIQLPOGUDBSU-UHFFFAOYSA-N | |
2,6-dimethylmorpholine | |
110862 | |
97% |
Safety and Handling
GHS H Statement
H318-H226-H312
Causes serious eye damage.
Flammable liquid and vapour.
Harmful in contact with skin.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H311-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 205-509-3
missing translation for 'rtecsNumber' : QE1750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only