Learn More
2,6-Dimethylmorpholine, 97%, mixture of isomers, Thermo Scientific Chemicals
91.70 € - 328.00 €
Chemical Identifiers
CAS | 141-91-3 |
---|---|
Molecular Formula | C6H13NO |
Molecular Weight (g/mol) | 115.17 |
InChI Key | HNVIQLPOGUDBSU-UHFFFAOYSA-N |
Synonym | morpholine, 2,6-dimethyl, 2,6-dimethyl morpholine, 2,6-dimethylmorfolin, 2,6-dimethylmorfolin czech, ccris 5910, trans-2,6-dimethylmorpholine, 2,6-dimethyl-2,3,5,6-tetrahydro-4h-1,4-oxazine, 2,6-dimethyl-morpholine, 2,6-dimethylmorpholine, cis + trans, acmc-209nod |
PubChem CID | 110862 |
IUPAC Name | 2,6-dimethylmorpholine |
SMILES | CC1CNCC(O1)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11916811
|
Thermo Scientific Acros
155340250 |
25 g |
91.70 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11926811
|
Thermo Scientific Acros
155341000 |
100 g |
328.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-91-3 | |
115.17 | |
morpholine, 2,6-dimethyl, 2,6-dimethyl morpholine, 2,6-dimethylmorfolin, 2,6-dimethylmorfolin czech, ccris 5910, trans-2,6-dimethylmorpholine, 2,6-dimethyl-2,3,5,6-tetrahydro-4h-1,4-oxazine, 2,6-dimethyl-morpholine, 2,6-dimethylmorpholine, cis + trans, acmc-209nod | |
2,6-dimethylmorpholine |
C6H13NO | |
HNVIQLPOGUDBSU-UHFFFAOYSA-N | |
110862 | |
CC1CNCC(O1)C |
Specifications
141-91-3 | |
0.9350g/mL | |
48°C | |
Glass bottle | |
1.447 | |
morpholine, 2,6-dimethyl, 2,6-dimethyl morpholine, 2,6-dimethylmorfolin, 2,6-dimethylmorfolin czech, ccris 5910, trans-2,6-dimethylmorpholine, 2,6-dimethyl-2,3,5,6-tetrahydro-4h-1,4-oxazine, 2,6-dimethyl-morpholine, 2,6-dimethylmorpholine, cis + trans, acmc-209nod | |
CC1CNCC(O1)C | |
115.17 | |
115.17 | |
2, 6-Dimethylmorpholine |
-85.0°C | |
147.0°C | |
96% min. (mixture of isomers) (GC) | |
C6H13NO | |
0.935 | |
HNVIQLPOGUDBSU-UHFFFAOYSA-N | |
2,6-dimethylmorpholine | |
110862 | |
97% |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-509-3
- 2,6-Dimethylmorpholine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.