missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-3-methoxyphenol, 97%, Thermo Scientific™

Product Code. 11396929
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886498-60-8
Molecular Formula C7H6F2O2
Molecular Weight (g/mol) 160.12
MDL Number MFCD04115907
InChI Key UOVAWRHEABQHGR-UHFFFAOYSA-N
Synonym 2,6-difluoro-3-methoxy-phenol, phenol,2,6-difluoro-3-methoxy, 2,6-bis fluoranyl-3-methoxy-phenol
PubChem CID 3682862
IUPAC Name 2,6-difluoro-3-methoxyphenol
SMILES COC1=C(C(=C(C=C1)F)O)F

Specifications

Melting Point 73°C to 76°C
Quantity 1 g
Formula Weight 160.12
Percent Purity 97%
Chemical Name or Material 2,6-Difluoro-3-methoxyphenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'