missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorothiobenzamide, 97%, Thermo Scientific™

Product Code. 11302529
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1918-13-4
Molecular Formula C7H5Cl2NS
Molecular Weight (g/mol) 206.08
MDL Number MFCD00053523
InChI Key KGKGSIUWJCAFPX-UHFFFAOYSA-N
Synonym 2,6-dichlorothiobenzamide, chlorthiamid, chlorthiamide, chlorothiamide, chlorthioamide, chlortiamid, prefix, dcbn, benzenecarbothioamide, 2,6-dichloro, 2,6-dichlorobenzothioamide
PubChem CID 2734819
ChEBI CHEBI:949
SMILES NC(=S)C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 149°C to 153°C
Flash Point >100°C (212°F)
Quantity 1 g
Beilstein 1910353
IUPAC Name 2,6-dichlorobenzene-1-carbothioamide
Formula Weight 206.09
Percent Purity 97%
Chemical Name or Material 2,6-Dichlorothiobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'