Learn More
2,6-Dichloroindophenol sodium salt hydrate, Thermo Scientific Chemicals
58.89 € - 668.33 €
Chemical Identifiers
CAS | 1266615-56-8 |
---|---|
Molecular Formula | C12H6Cl2NNaO2 |
Molecular Weight (g/mol) | 290.07 |
MDL Number | MFCD00150014 |
InChI Key | FHLDWQLHDYCXKI-UHFFFAOYSA-M |
Synonym | 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt |
PubChem CID | 23697355 |
ChEBI | CHEBI:948 |
IUPAC Name | sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate |
SMILES | [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449342
|
Thermo Scientific Alfa Aesar
A10107.06 |
5 g |
58.89 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459342
|
Thermo Scientific Alfa Aesar
A10107.14 |
25 g |
201.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11469342
|
Thermo Scientific Alfa Aesar
A10107.22 |
100 g |
668.33 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
1266615-56-8 | |
290.07 | |
FHLDWQLHDYCXKI-UHFFFAOYSA-M | |
23697355 | |
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate |
C12H6Cl2NNaO2 | |
MFCD00150014 | |
2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt | |
CHEBI:948 | |
[Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1 |
Specifications
1266615-56-8 | |
C12H6Cl2NNaO2 | |
3641229 | |
14,3068 | |
Soluble in water. | |
[Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1 | |
290.07 | |
CHEBI:948 | |
2,6-Dichloroindophenol sodium salt hydrate |
>300°C | |
MFCD00150014 | |
Light sensitive | |
2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt | |
FHLDWQLHDYCXKI-UHFFFAOYSA-M | |
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate | |
23697355 | |
290.08 (Anhydrous) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
H500
missing translation for 'einecsNumber' : 210-640-4
missing translation for 'rtecsNumber' : GU5495000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 2,6-Dichloroindophenol sodium salt hydrate
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.