missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol, sodium salt hydrate, 90+%, Thermo Scientific Chemicals

52.06 € - 287.43 €

Chemical Identifiers

CAS 1266615-56-8
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key CVSUAFOWIXUYQA-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt dihydrate, 2,6-dichloro-4-4-hydroxyphenyl azamethylene cyclohexa-2,5-dien-1-one, oxamet hane, oxamethane, sodium salt, c12h6cl2no2.na.2h2o, 2,6-dichlorophenol-indophenol sodium salt dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino phenolate dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino benzenolate dihydrate
PubChem CID 23696612
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate;dihydrate
SMILES [Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
13565020
View Documents
Thermo Scientific Acros
450700050
5 g Glass bottle
52.06 €
5g
Please to purchase this item. Need a web account? Register with us today!
13505050
View Documents
Thermo Scientific Acros
450700250
25 g Glass bottle
114.57 €
25g
Please to purchase this item. Need a web account? Register with us today!
13515050
View Documents
Thermo Scientific Acros
450701000
100 g Glass bottle
287.43 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1266615-56-8
290.07
CVSUAFOWIXUYQA-UHFFFAOYSA-M
23696612
[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
C12H6Cl2NNaO2
MFCD00150014
2,6-dichloroindophenol sodium salt dihydrate, 2,6-dichloro-4-4-hydroxyphenyl azamethylene cyclohexa-2,5-dien-1-one, oxamet hane, oxamethane, sodium salt, c12h6cl2no2.na.2h2o, 2,6-dichlorophenol-indophenol sodium salt dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino phenolate dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino benzenolate dihydrate
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate;dihydrate

Specifications

1266615-56-8
C12H6Cl2NNaO2
2,6-dichloroindophenol sodium salt dihydrate, 2,6-dichloro-4-4-hydroxyphenyl azamethylene cyclohexa-2,5-dien-1-one, oxamet hane, oxamethane, sodium salt, c12h6cl2no2.na.2h2o, 2,6-dichlorophenol-indophenol sodium salt dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino phenolate dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino benzenolate dihydrate
CVSUAFOWIXUYQA-UHFFFAOYSA-M
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate;dihydrate
23696612
90+%
90+%
MFCD00150014
Solubility in water: soluble. Other solubilities: freely soluble in alcohol
[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
290.07
290.07
2, 6-Dichloroindophenol, sodium salt hydrate
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.