Learn More
2,6-Dichlorobenzonitrile, 98%, Thermo Scientific Chemicals
20.60 € - 94.67 €
Chemical Identifiers
CAS | 1194-65-6 |
---|---|
Molecular Formula | C7H3Cl2N |
Molecular Weight (g/mol) | 172.008 |
MDL Number | MFCD00001781 |
InChI Key | YOYAIZYFCNQIRF-UHFFFAOYSA-N |
Synonym | dichlobenil, dichlobanil, casoron, prefix d, benzonitrile, 2,6-dichloro, casaron, du-sprex, decabane, surfassol, casoron 133 |
PubChem CID | 3031 |
ChEBI | CHEBI:943 |
IUPAC Name | 2,6-dichlorobenzonitrile |
SMILES | C1=CC(=C(C(=C1)Cl)C#N)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11464060
|
Thermo Scientific Alfa Aesar
A14986.09 |
10 g |
20.60 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11417207
|
Thermo Scientific Alfa Aesar
A14986.14 |
25 g |
32.96 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427207
|
Thermo Scientific Alfa Aesar
A14986.22 |
100 g |
94.67 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-dichlorobenzonitrile in which Q-amino-G- chlorobenzonitrile is used as starting material. And it is used for the synthesis of diclofenac sodium oxacillin and guanabenz acetate.
Solubility
Soluble in water (0.018g/L).
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
1194-65-6 | |
172.008 | |
YOYAIZYFCNQIRF-UHFFFAOYSA-N | |
3031 | |
2,6-dichlorobenzonitrile |
C7H3Cl2N | |
MFCD00001781 | |
dichlobenil, dichlobanil, casoron, prefix d, benzonitrile, 2,6-dichloro, casaron, du-sprex, decabane, surfassol, casoron 133 | |
CHEBI:943 | |
C1=CC(=C(C(=C1)Cl)C#N)Cl |
Specifications
1194-65-6 | |
270°C | |
C7H3Cl2N | |
UN3077 | |
14,3042 | |
YOYAIZYFCNQIRF-UHFFFAOYSA-N | |
2,6-dichlorobenzonitrile | |
3031 | |
172.01 | |
2,6-Dichlorobenzonitrile |
143°C to 147°C | |
Aromatic | |
MFCD00001781 | |
1909167 | |
dichlobenil, dichlobanil, casoron, prefix d, benzonitrile, 2,6-dichloro, casaron, du-sprex, decabane, surfassol, casoron 133 | |
C1=CC(=C(C(=C1)Cl)C#N)Cl | |
172.008 | |
CHEBI:943 | |
98% |
Safety and Handling
GHS H Statement
H312
Harmful in contact with skin.
GHS P Statement
P280-P312-P363-P322-P302+P352-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'rtecsNumber' : DI3500000
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.