Learn More
2,6-Dibromo-N-chloro-p-benzoquinoneimine, 97%, Thermo Scientific Chemicals
158.00 € - 594.96 €
Chemical Identifiers
CAS | 537-45-1 |
---|---|
Molecular Formula | C6H2Br2ClNO |
Molecular Weight (g/mol) | 299.34 |
InChI Key | JYWKEVKEKOTYEX-UHFFFAOYSA-N |
Synonym | 2,6-dibromoquinone-4-chloroimide, 2,6-dibromo-4-chloroimino cyclohexa-2,5-dienone, bqc reagent, 2,6-dibromoquinone-4-chlorimide, 2,6-dibromoquinone chlorimide, 2,6-dibromoquinone chloroimide, 2,6-dibromoquinone chloroimine, n-chloro-2,6-dibromoquinoneimine, 2,5-cyclohexadien-1-one, 2,6-dibromo-4-chloroimino, 2,6-dibromo-p-benzoquinone-4-chlorimine |
PubChem CID | 10835 |
IUPAC Name | 2,6-dibromo-4-chloroiminocyclohexa-2,5-dien-1-one |
SMILES | C1=C(C(=O)C(=CC1=NCl)Br)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10341391
|
Thermo Scientific Acros
406470050 |
5 g | Glass bottle |
158.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11925601
|
Thermo Scientific Acros
406470250 |
25 g | Glass bottle |
594.96 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
537-45-1 | |
299.34 | |
2,6-dibromoquinone-4-chloroimide, 2,6-dibromo-4-chloroimino cyclohexa-2,5-dienone, bqc reagent, 2,6-dibromoquinone-4-chlorimide, 2,6-dibromoquinone chlorimide, 2,6-dibromoquinone chloroimide, 2,6-dibromoquinone chloroimine, n-chloro-2,6-dibromoquinoneimine, 2,5-cyclohexadien-1-one, 2,6-dibromo-4-chloroimino, 2,6-dibromo-p-benzoquinone-4-chlorimine | |
2,6-dibromo-4-chloroiminocyclohexa-2,5-dien-1-one |
C6H2Br2ClNO | |
JYWKEVKEKOTYEX-UHFFFAOYSA-N | |
10835 | |
C1=C(C(=O)C(=CC1=NCl)Br)Br |
Specifications
537-45-1 | |
100.0 | |
Yellow to Ochre | |
Authentic | |
C6H2Br2ClNO | |
1.81 | |
JYWKEVKEKOTYEX-UHFFFAOYSA-N | |
2,6-dibromo-4-chloroiminocyclohexa-2,5-dien-1-one | |
10835 | |
97% | |
2, 6-Dibromo-N-chloro-p-benzoquinoneimine |
96.0 | |
76.0°C to 83.0°C | |
1.8100g/mL | |
96% min. (Iodometry) | |
15,3031 | |
2,6-dibromoquinone-4-chloroimide, 2,6-dibromo-4-chloroimino cyclohexa-2,5-dienone, bqc reagent, 2,6-dibromoquinone-4-chlorimide, 2,6-dibromoquinone chlorimide, 2,6-dibromoquinone chloroimide, 2,6-dibromoquinone chloroimine, n-chloro-2,6-dibromoquinoneimine, 2,5-cyclohexadien-1-one, 2,6-dibromo-4-chloroimino, 2,6-dibromo-p-benzoquinone-4-chlorimine | |
C1=C(C(=O)C(=CC1=NCl)Br)Br | |
299.34 | |
299.34 | |
Powder or Fine Crystalline Needles |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Heating may cause a fire.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-667-1
Recommended Storage : Product may discolor to orange-brown on storage
- 2,6-Dibromo-N-chloro-p-benzoquinoneimine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Self-reaC & Dtive substanC & De or mixture Type C & D
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H242-Heating may cause a fire.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only