Learn More
2,5-Dimethyl-p-benzoquinone, 99%, Thermo Scientific Chemicals
78.00 € - 263.00 €
Chemical Identifiers
CAS | 137-18-8 |
---|---|
Molecular Formula | C8H8O2 |
Molecular Weight (g/mol) | 136.15 |
MDL Number | MFCD00041737 |
InChI Key | MYKLQMNSFPAPLZ-UHFFFAOYSA-N |
Synonym | 2,5-dimethyl-1,4-benzoquinone, phlorone, p-xyloquinone, 2,5-dimethyl-p-benzoquinone, 2,5-xyloquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dimethyl, 2,5-dimethylquinone, floron, 3,6-dimethyl-p-benzoquinone, floron czech |
PubChem CID | 8718 |
IUPAC Name | 2,5-dimethylcyclohexa-2,5-diene-1,4-dione |
SMILES | CC1=CC(=O)C(=CC1=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10055790
|
Thermo Scientific Acros
408230050 |
5 g | Glass bottle |
78.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10422815
|
Thermo Scientific Acros
408230250 |
25 g | Glass bottle |
263.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
137-18-8 | |
136.15 | |
MYKLQMNSFPAPLZ-UHFFFAOYSA-N | |
8718 | |
CC1=CC(=O)C(=CC1=O)C |
C8H8O2 | |
MFCD00041737 | |
2,5-dimethyl-1,4-benzoquinone, phlorone, p-xyloquinone, 2,5-dimethyl-p-benzoquinone, 2,5-xyloquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dimethyl, 2,5-dimethylquinone, floron, 3,6-dimethyl-p-benzoquinone, floron czech | |
2,5-dimethylcyclohexa-2,5-diene-1,4-dione |
Specifications
137-18-8 | |
100.0 | |
Yellow to Brown | |
98.5% min. (GC) | |
MFCD00041737 | |
MYKLQMNSFPAPLZ-UHFFFAOYSA-N | |
2,5-dimethylcyclohexa-2,5-diene-1,4-dione | |
8718 | |
99% | |
2, 5-Dimethyl-p-benzoquinone |
98.5 | |
123°C to 126°C | |
Authentic | |
C8H8O2 | |
2,5-dimethyl-1,4-benzoquinone, phlorone, p-xyloquinone, 2,5-dimethyl-p-benzoquinone, 2,5-xyloquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dimethyl, 2,5-dimethylquinone, floron, 3,6-dimethyl-p-benzoquinone, floron czech | |
CC1=CC(=O)C(=CC1=O)C | |
136.15 | |
136.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep a
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-283-6
- 2,5-Dimethyl-p-benzoquinone
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only