missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dihydroxyacetophenone, 98+%, Thermo Scientific Chemicals

Product Code. 11424833
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490-78-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002343
InChI Key WLDWSGZHNBANIO-UHFFFAOYSA-N
Synonym 2',5'-dihydroxyacetophenone, 1-2,5-dihydroxyphenyl ethanone, 2-acetylhydroquinone, 2,5-dihydroxyacetophenone, quinacetophenone, acetylhydroquinone, ethanone, 1-2,5-dihydroxyphenyl, 1-2,5-dihydroxyphenyl ethan-1-one, acetylquinol, acetophenone, 2',5'-dihydroxy
PubChem CID 10279
IUPAC Name 1-(2,5-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)O)O

Specifications

Melting Point 204°C to 208°C
Quantity 25 g
Beilstein 637903
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 2',5'-Dihydroxyacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'