missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11438116
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21663-61-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00019741
InChI Key LNGWRTKJZCBXGT-UHFFFAOYSA-N
Synonym benzonitrile, 2,5-dichloro, 2,5-dichloro-benzonitrile, 2,5-dichlorobenzonitrite, 2,5-dichlorobenzenecarbonitrile, pubchem3687, acmc-209fn5, ksc490q4h, 2,5-dichlorobenzonitrile, attercop-chm at133057
PubChem CID 89000
IUPAC Name 2,5-dichlorobenzonitrile
SMILES C1=CC(=C(C=C1Cl)C#N)Cl

Specifications

Melting Point 128°C to 131°C
Quantity 100 g
UN Number UN3439
Beilstein 2045293
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorobenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'