missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-p-benzoquinone, 98%, Thermo Scientific Chemicals

24.41 € - 231.15 €

Chemical Identifiers

CAS 615-93-0
Molecular Formula C6H2Cl2O2
Molecular Weight (g/mol) 176.99
InChI Key LNXVNZRYYHFMEY-UHFFFAOYSA-N
Synonym 2,5-dichloro-1,4-benzoquinone, 2,5-dichloro-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro, 2,5-dichlorobenzo-1,4-quinone, p-benzoquinone, 2,5-dichloro, 2,5-dichloro-p-quinone, acmc-1bhnt, 2,5-dichlorobenzoquinone, p-benzoquinone,5-dichloro, 2,5-dichoro-p-benzoquinone
PubChem CID 12011
IUPAC Name 2,5-dichlorocyclohexa-2,5-diene-1,4-dione
SMILES C1=C(C(=O)C=C(C1=O)Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15515444
missing translation for 'viewDocuments'
Thermo Scientific Acros
406730010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15760869
missing translation for 'viewDocuments'
Thermo Scientific Acros
406730050
5 g
79.90 €
5g
Please to purchase this item. Need a web account? Register with us today!
15770869
missing translation for 'viewDocuments'
Thermo Scientific Acros
406730250
25 g
231.15 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

615-93-0
176.99
2,5-dichloro-1,4-benzoquinone, 2,5-dichloro-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro, 2,5-dichlorobenzo-1,4-quinone, p-benzoquinone, 2,5-dichloro, 2,5-dichloro-p-quinone, acmc-1bhnt, 2,5-dichlorobenzoquinone, p-benzoquinone,5-dichloro, 2,5-dichoro-p-benzoquinone
2,5-dichlorocyclohexa-2,5-diene-1,4-dione
C6H2Cl2O2
LNXVNZRYYHFMEY-UHFFFAOYSA-N
12011
C1=C(C(=O)C=C(C1=O)Cl)Cl

Specifications

615-93-0
Authentic
Glass Bottle
2,5-dichloro-1,4-benzoquinone, 2,5-dichloro-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro, 2,5-dichlorobenzo-1,4-quinone, p-benzoquinone, 2,5-dichloro, 2,5-dichloro-p-quinone, acmc-1bhnt, 2,5-dichlorobenzoquinone, p-benzoquinone,5-dichloro, 2,5-dichoro-p-benzoquinone
C1=C(C(=O)C=C(C1=O)Cl)Cl
176.99
176.99
2,5-Dichloro-p-benzoquinone
159.0°C to 161.0°C
97.5% min. (GC)
C6H2Cl2O2
LNXVNZRYYHFMEY-UHFFFAOYSA-N
2,5-dichlorocyclohexa-2,5-diene-1,4-dione
12011
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 210-453-8

Product Identifier
  • 2,5-Dichloro-p-benzoquinone, 98%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Certificates

RUO – Research Use Only