missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pyridinedicarboxylic acid hydrate, 99+%, Thermo Scientific Chemicals

19.20 € - 165.83 €

Chemical Identifiers

CAS 499-80-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00149413,MFCD00006296
InChI Key MJIVRKPEXXHNJT-UHFFFAOYSA-N
Synonym 2,4-pyridinedicarboxylic acid, lutidinic acid, 2,4-lutidinic acid, 2,4-pyridinedicarboxylicacid, hydrate 1:1, 4nrq, pd2, 4-carboxypicolinic acid, pubchem16314, pyridine carboxylate, 6a, acmc-1bnb9
PubChem CID 10365
ChEBI CHEBI:44737
IUPAC Name pyridine-2,4-dicarboxylic acid
SMILES OC(=O)C1=CC=NC(=C1)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10720595
View Documents Promotion Details
Thermo Scientific Acros
131860010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10376870
View Documents Promotion Details
Thermo Scientific Acros
131860050
5 g Glass bottle
50.85 €
5g
Please to purchase this item. Need a web account? Register with us today!
10540994
View Documents Promotion Details
Thermo Scientific Acros
131860250
25 g Glass bottle
165.83 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

499-80-9
167.12
MJIVRKPEXXHNJT-UHFFFAOYSA-N
10365
pyridine-2,4-dicarboxylic acid
C7H5NO4
MFCD00149413,MFCD00006296
2,4-pyridinedicarboxylic acid, lutidinic acid, 2,4-lutidinic acid, 2,4-pyridinedicarboxylicacid, hydrate 1:1, 4nrq, pd2, 4-carboxypicolinic acid, pubchem16314, pyridine carboxylate, 6a, acmc-1bnb9
CHEBI:44737
OC(=O)C1=CC=NC(=C1)C(O)=O

Specifications

499-80-9
100.0
White
99+%
(C5H3N)(COOH)2·xH2O
22, 153
Solubility in water: insoluble
OC(=O)C1=CC=NC(=C1)C(O)=O
167.12
CHEBI:44737
99+%
2, 4-Pyridinedicarboxylic acid hydrate, 99+%
99.0
243°C to 246°C
Authentic
C7H5NO4
MFCD00149413,MFCD00006296
2,4-pyridinedicarboxylic acid, lutidinic acid, 2,4-lutidinic acid, 2,4-pyridinedicarboxylicacid, hydrate 1:1, 4nrq, pd2, 4-carboxypicolinic acid, pubchem16314, pyridine carboxylate, 6a, acmc-1bnb9
MJIVRKPEXXHNJT-UHFFFAOYSA-N
pyridine-2,4-dicarboxylic acid
10365
167.12
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

missing translation for 'einecsNumber' : 207-892-2

missing translation for 'tsca' : TSCA

Product Identifier
  • 2,4-Pyridinedicarboxylic acid hydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only