missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Piperidinyloxy)-N-propylacetamide, 95%, Thermo Scientific™

Product Code. 11404910
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902836-22-0
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD08064265
InChI Key TZQLDIRIOACOLB-UHFFFAOYSA-N
Synonym 2-4-piperidinyloxy-n-propylacetamide, 2-piperidin-4-yloxy-n-propylacetamide, 2-4-piperidyloxy-n-propylacetamide, 2-piperidin-4-yl-oxy-n-propylacetamide, 2-piperidin-4-yl oxy-n-propylacetamide
PubChem CID 24208844
IUPAC Name 2-piperidin-4-yloxy-N-propylacetamide
SMILES CCCNC(=O)COC1CCNCC1

Specifications

Quantity 1 g
Formula Weight 200.28
Percent Purity 95%
Chemical Name or Material 2-(4-Piperidinyloxy)-N-propylacetamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'