missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Piperidinyloxy)benzonitrile, 98%, Thermo Scientific™

Product Code. 11489710
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 900572-37-4
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.26
MDL Number MFCD08061090
InChI Key OIFRRCRLKBGBKR-UHFFFAOYSA-N
Synonym 2-4-piperidinyloxy benzonitrile, 2-piperidin-4-yloxy benzonitrile, acmc-20aoor, 2-4-piperidyloxy benzenecarbonitrile, 2-piperidin-4-yl oxy benzonitrile, benzonitrile, 2-4-piperidinyloxy, 2-piperidin-4-yl-oxy-benzonitrile
PubChem CID 16732591
IUPAC Name 2-piperidin-4-yloxybenzonitrile
SMILES N#CC1=CC=CC=C1OC1CCNCC1

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 202.26
Percent Purity 98%
Chemical Name or Material 2-(4-Piperidinyloxy)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'