missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Morpholinyl)thiazole-5-carboxaldehyde, 97%, Thermo Scientific™

66.10 € - 208.00 €

Chemical Identifiers

CAS 1011-41-2
Molecular Formula C8H10N2O2S
Molecular Weight (g/mol) 198.24
MDL Number MFCD01568825
InChI Key VDZWHWVAMDQEBT-UHFFFAOYSA-N
Synonym 2-morpholino-1,3-thiazole-5-carbaldehyde, 2-morpholin-4-yl-1,3-thiazole-5-carbaldehyde, 2-morpholin-4-yl-thiazole-5-carbaldehyde, 2-morpholinothiazole-5-carbaldehyde, acmc-20anpj, morpholinothiazolecarbaldehyde, 5-formyl-2-morpholin-4-yl-1,3-thiazole, 5-thiazolecarboxaldehyde,2-4-morpholinyl, 2-4-morpholinyl-1,3-thiazole-5-carbaldehyde
PubChem CID 1479770
IUPAC Name 2-morpholin-4-yl-1,3-thiazole-5-carbaldehyde
SMILES O=CC1=CN=C(S1)N1CCOCC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11375509
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50463.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11385509
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50463.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1011-41-2
198.24
VDZWHWVAMDQEBT-UHFFFAOYSA-N
1479770
O=CC1=CN=C(S1)N1CCOCC1
C8H10N2O2S
MFCD01568825
2-morpholino-1,3-thiazole-5-carbaldehyde, 2-morpholin-4-yl-1,3-thiazole-5-carbaldehyde, 2-morpholin-4-yl-thiazole-5-carbaldehyde, 2-morpholinothiazole-5-carbaldehyde, acmc-20anpj, morpholinothiazolecarbaldehyde, 5-formyl-2-morpholin-4-yl-1,3-thiazole, 5-thiazolecarboxaldehyde,2-4-morpholinyl, 2-4-morpholinyl-1,3-thiazole-5-carbaldehyde
2-morpholin-4-yl-1,3-thiazole-5-carbaldehyde

Specifications

1011-41-2
MFCD01568825
VDZWHWVAMDQEBT-UHFFFAOYSA-N
2-morpholin-4-yl-1,3-thiazole-5-carbaldehyde
1479770
97%
C8H10N2O2S
2-morpholino-1,3-thiazole-5-carbaldehyde, 2-morpholin-4-yl-1,3-thiazole-5-carbaldehyde, 2-morpholin-4-yl-thiazole-5-carbaldehyde, 2-morpholinothiazole-5-carbaldehyde, acmc-20anpj, morpholinothiazolecarbaldehyde, 5-formyl-2-morpholin-4-yl-1,3-thiazole, 5-thiazolecarboxaldehyde,2-4-morpholinyl, 2-4-morpholinyl-1,3-thiazole-5-carbaldehyde
O=CC1=CN=C(S1)N1CCOCC1
198.24
198.25
2-(4-Morpholinyl)thiazole-5-carboxaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications