Learn More
2-(4-Morpholinyl)aniline, 98%, Thermo Scientific Chemicals
78.90 € - 270.00 €
Chemical Identifiers
CAS | 5585-33-1 |
---|---|
Molecular Formula | C10H14N2O |
Molecular Weight (g/mol) | 178.235 |
MDL Number | MFCD00047408 |
InChI Key | QKWLVAYDAHQMLG-UHFFFAOYSA-N |
Synonym | 2-morpholinoaniline, 2-morpholin-4-yl aniline, 2-morpholin-4-yl-phenylamine, 2-4-morpholino aniline, 4-2-aminophenyl morpholine, 2-4-morpholinyl aniline, 2-morpholin-4-ylphenylamine, enamine_000782, acmc-1ap2s |
PubChem CID | 735756 |
IUPAC Name | 2-morpholin-4-ylaniline |
SMILES | C1COCCN1C2=CC=CC=C2N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337535
|
Thermo Scientific Alfa Aesar
L05660.06 |
5 g |
78.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347535
|
Thermo Scientific Alfa Aesar
L05660.14 |
25 g |
270.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5585-33-1 | |
178.235 | |
QKWLVAYDAHQMLG-UHFFFAOYSA-N | |
735756 | |
C1COCCN1C2=CC=CC=C2N |
C10H14N2O | |
MFCD00047408 | |
2-morpholinoaniline, 2-morpholin-4-yl aniline, 2-morpholin-4-yl-phenylamine, 2-4-morpholino aniline, 4-2-aminophenyl morpholine, 2-4-morpholinyl aniline, 2-morpholin-4-ylphenylamine, enamine_000782, acmc-1ap2s | |
2-morpholin-4-ylaniline |
Especificaciones
5585-33-1 | |
C10H14N2O | |
UN2811 | |
2-morpholinoaniline, 2-morpholin-4-yl aniline, 2-morpholin-4-yl-phenylamine, 2-4-morpholino aniline, 4-2-aminophenyl morpholine, 2-4-morpholinyl aniline, 2-morpholin-4-ylphenylamine, enamine_000782, acmc-1ap2s | |
C1COCCN1C2=CC=CC=C2N | |
178.235 | |
178.24 | |
2-(4-Morpholinyl)aniline |
95°C to 98°C | |
MFCD00047408 | |
163926 | |
QKWLVAYDAHQMLG-UHFFFAOYSA-N | |
2-morpholin-4-ylaniline | |
735756 | |
98% |
Seguridad y manipulación
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only