missing translation for 'onlineSavingsMsg'
Learn More

2-[(4-Fluorophenyl)dimethylsilyl]benzyl alcohol, 95%, Thermo Scientific™

39.00 € - 149.00 €

Chemical Identifiers

CAS 853955-70-1
Molecular Formula C15H17FOSi
Molecular Weight (g/mol) 260.383
MDL Number MFCD16251550
InChI Key RKOHTBFNQRVSEP-UHFFFAOYSA-N
Synonym 2-4-fluorophenyl dimethylsilyl benzyl alcohol, 2-4-fluorophenyl dimethylsilyl phenyl methanol, 2-4-fluoro-phenyl-dimethyl-silanyl-phenyl-methanol, amtsi119, 2-4-fluorophenyl dimethylsilyl benzene-1-methanol, 2-4-fluorophenyl dimethyl silyl phenyl methanol
PubChem CID 11989534
IUPAC Name [2-[(4-fluorophenyl)-dimethylsilyl]phenyl]methanol
SMILES C[Si](C)(C1=CC=C(C=C1)F)C2=CC=CC=C2CO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15499295
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51735.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15409305
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H51735.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

853955-70-1
260.383
RKOHTBFNQRVSEP-UHFFFAOYSA-N
11989534
C[Si](C)(C1=CC=C(C=C1)F)C2=CC=CC=C2CO
C15H17FOSi
MFCD16251550
2-4-fluorophenyl dimethylsilyl benzyl alcohol, 2-4-fluorophenyl dimethylsilyl phenyl methanol, 2-4-fluoro-phenyl-dimethyl-silanyl-phenyl-methanol, amtsi119, 2-4-fluorophenyl dimethylsilyl benzene-1-methanol, 2-4-fluorophenyl dimethyl silyl phenyl methanol
[2-[(4-fluorophenyl)-dimethylsilyl]phenyl]methanol

Specifications

853955-70-1
MFCD16251550
RKOHTBFNQRVSEP-UHFFFAOYSA-N
[2-[(4-fluorophenyl)-dimethylsilyl]phenyl]methanol
11989534
95%
C15H17FOSi
2-4-fluorophenyl dimethylsilyl benzyl alcohol, 2-4-fluorophenyl dimethylsilyl phenyl methanol, 2-4-fluoro-phenyl-dimethyl-silanyl-phenyl-methanol, amtsi119, 2-4-fluorophenyl dimethylsilyl benzene-1-methanol, 2-4-fluorophenyl dimethyl silyl phenyl methanol
C[Si](C)(C1=CC=C(C=C1)F)C2=CC=CC=C2CO
260.383
260.38
2-[(4-Fluorophenyl)dimethylsilyl]benzyl alcohol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications