missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dimethylacetanilide, 98%, Thermo Scientific™

Product Code. 15401928
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2050-43-3
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00026125
InChI Key PJEIIBXJUDOMAN-UHFFFAOYSA-N
Synonym n-2,4-dimethylphenyl acetamide, 2',4'-dimethylacetanilide, 2,4-dimethylacetanilide, 4-acetamido-m-xylene, 2',4'-acetoxylidine, 2',4'-acetoxylidide, acetamide, n-2,4-dimethylphenyl, acetanilide, 2',4'-dimethyl, n-acetyl-2,4-xylidine, unii-3u0w52hz1h
PubChem CID 16303
IUPAC Name N-(2,4-dimethylphenyl)acetamide
SMILES CC1=CC(=C(C=C1)NC(=O)C)C

Specifications

Melting Point 129°C to 132°C
Color White
Quantity 25 g
Formula Weight 163.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2',4'-Dimethylacetanilide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'