missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 11475807
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133730-34-4
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.982
MDL Number MFCD01074590
InChI Key SQTUYFKNCCBFRR-UHFFFAOYSA-N
Synonym 2,4-dimethoxybenzeneboronic acid, 2,4-dimethoxyphenyl boronic acid, 2,4-dimethoxyphenyl boranediol, boronic acid, 2,4-dimethoxyphenyl, 2,4-dimethoxybenzene boronic acid, boronic acid, b-2,4-dimethoxyphenyl, pubchem1823, acmc-209btd, ksc174i6j, 2,4-dimethoxyphenylboronicacid
PubChem CID 2734341
IUPAC Name (2,4-dimethoxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)OC)OC)(O)O

Specifications

Melting Point 115°C to 120°C
Quantity 5 g
Beilstein 4744213
Formula Weight 181.98
Percent Purity 98%
Chemical Name or Material 2,4-Dimethoxybenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'