missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyaniline, 97%, Thermo Scientific™

21.70 € - 56.80 €

Chemical Identifiers

CAS 2735-04-8
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008371
InChI Key GEQNZVKIDIPGCO-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dimethoxy, aniline, 2,4-dimethoxy, 2,4-dimethoxyphenylamine, unii-i079354gsi, benzenamine, 2,4-dimethoxy-, hydrochloride, 2,4-domethoxyaniline, 2,4-dimethoxy-anilin, 2, 4-dimethoxyaniline, 2,4 dimethoxy aniline, 2,4-dimethoxy aniline
PubChem CID 17652
IUPAC Name 2,4-dimethoxyaniline
SMILES COC1=CC(=C(C=C1)N)OC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10606112
View Documents
Thermo Scientific Acros
115350050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10261233
View Documents
Thermo Scientific Acros
115352500
250 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2735-04-8
MFCD00008371
benzenamine, 2,4-dimethoxy, aniline, 2,4-dimethoxy, 2,4-dimethoxyphenylamine, unii-i079354gsi, benzenamine, 2,4-dimethoxy-, hydrochloride, 2,4-domethoxyaniline, 2,4-dimethoxy-anilin, 2, 4-dimethoxyaniline, 2,4 dimethoxy aniline, 2,4-dimethoxy aniline
2,4-dimethoxyaniline
153.18
GEQNZVKIDIPGCO-UHFFFAOYSA-N
17652
COC1=CC(=C(C=C1)N)OC

Specifications

2735-04-8
100.0
Brown
Authentic
C8H11NO2
MFCD00008371
benzenamine, 2,4-dimethoxy, aniline, 2,4-dimethoxy, 2,4-dimethoxyphenylamine, unii-i079354gsi, benzenamine, 2,4-dimethoxy-, hydrochloride, 2,4-domethoxyaniline, 2,4-dimethoxy-anilin, 2, 4-dimethoxyaniline, 2,4 dimethoxy aniline, 2,4-dimethoxy aniline
GEQNZVKIDIPGCO-UHFFFAOYSA-N
2,4-dimethoxyaniline
17652
97%
2, 4-Dimethoxyaniline, 97%
96.0
34°C to 37°C
>110°C
96% min. (GC)
(CH3O)2C6H3NH2
13, 784
Solubility in water: 7g/L (20°C)
COC1=CC(=C(C=C1)N)OC
153.18
153.18
Crystalline Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 220-355-7

missing translation for 'rtecsNumber' : BX4200000

missing translation for 'tsca' : TSCA

Product Identifier
  • 2,4-Dimethoxyaniline

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.