missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Difluoroacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11465453
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 364-83-0
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.132
MDL Number MFCD00151261
InChI Key QEWHNJPLPZOEKU-UHFFFAOYSA-N
Synonym 2',4'-difluoroacetophenone, 1-2,4-difluorophenyl ethanone, 2,4-difluoroacetophenone, 1-2,4-difluorophenyl ethan-1-one, ethanone, 1-2,4-difluorophenyl, 1-2,4-difluoro-phenyl-ethanone, 1-acetyl-2,4-difluorobenzene, acetophenone, 2',4'-difluoro, 1-2,4-difluorphenyl ethanon, pubchem4213
PubChem CID 67770
IUPAC Name 1-(2,4-difluorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)F)F

Specifications

Density 1.232
Boiling Point 80°C to 81°C (25 mmHg)
Flash Point 66°C (150°F)
Odor Characteristic
Refractive Index 1.488
Quantity 5 g
Beilstein 1940741
Formula Weight 156.13
Percent Purity 98%
Chemical Name or Material 2',4'-Difluoroacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.