missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-N-methylaniline, 98%, Thermo Scientific™

Product Code. 11482438
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138564-16-6
Molecular Formula C7H7F2N
Molecular Weight (g/mol) 143.14
MDL Number MFCD00042472
InChI Key RPGDVPAENGWKAO-UHFFFAOYSA-N
Synonym n-methyl-2,4-difluoroaniline, n1-methyl-2,4-difluoroaniline, benzenamine,2,4-difluoro-n-methyl, benzenamine, 2,4-difluoro-n-methyl, difluoronmethylaniline, pubchem10041, acmc-20aod4, 2,4-difluoro-n-methylaniline, 2,4-difluoro-phenyl-methyl-amine, 2,4-bis fluoranyl-n-methyl-aniline
PubChem CID 2737023
IUPAC Name 2,4-difluoro-N-methylaniline
SMILES CNC1=CC=C(F)C=C1F

Specifications

Boiling Point 94°C to 96°C (15 mmHg)
Refractive Index 1.5045
Quantity 25 g
UN Number UN2810
Beilstein 7019861
Formula Weight 143.14
Percent Purity 98%
Chemical Name or Material 2,4-Difluoro-N-methylaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'