Learn More
2,4-Difluoro-5-nitrophenol, 98%, Thermo Scientific Chemicals
88.24 € - 284.42 €
Chemical Identifiers
CAS | 113512-57-5 |
---|---|
Molecular Formula | C6H3F2NO3 |
Molecular Weight (g/mol) | 175.091 |
MDL Number | MFCD08443942 |
InChI Key | SMRYCTJAGPDVEH-UHFFFAOYSA-N |
Synonym | 2,4-difluoro-5-nitrobenzenol, phenol, 2,4-difluoro-5-nitro, difluoronitrobenzenol, acmc-209wyr, 2,4-difluoro-5-hydroxynitrobenzene |
PubChem CID | 10921024 |
IUPAC Name | 2,4-difluoro-5-nitrophenol |
SMILES | C1=C(C(=CC(=C1O)F)F)[N+](=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15444757
|
Thermo Scientific Alfa Aesar
H32081.06 |
5 g |
88.24 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15454757
|
Thermo Scientific Alfa Aesar
H32081.14 |
25 g |
284.42 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,4-Difluoro-5-nitrophenol may be used in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Difluoro-5-nitrophenol may be used in chemical synthesis.
Solubility
Sparingly soluble in water but readily soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
113512-57-5 | |
175.091 | |
SMRYCTJAGPDVEH-UHFFFAOYSA-N | |
10921024 | |
C1=C(C(=CC(=C1O)F)F)[N+](=O)[O-] |
C6H3F2NO3 | |
MFCD08443942 | |
2,4-difluoro-5-nitrobenzenol, phenol, 2,4-difluoro-5-nitro, difluoronitrobenzenol, acmc-209wyr, 2,4-difluoro-5-hydroxynitrobenzene | |
2,4-difluoro-5-nitrophenol |
Specifications
113512-57-5 | |
C6H3F2NO3 | |
2,4-difluoro-5-nitrobenzenol, phenol, 2,4-difluoro-5-nitro, difluoronitrobenzenol, acmc-209wyr, 2,4-difluoro-5-hydroxynitrobenzene | |
SMRYCTJAGPDVEH-UHFFFAOYSA-N | |
2,4-difluoro-5-nitrophenol | |
10921024 | |
98% |
112°C to 120°C | |
MFCD08443942 | |
Sparingly soluble in water but readily soluble in organic solvents. | |
C1=C(C(=CC(=C1O)F)F)[N+](=O)[O-] | |
175.091 | |
175.09 | |
2,4-Difluoro-5-nitrophenol |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.