missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methoxypyrimidine, 97%, Thermo Scientific Chemicals

103.52 € - 923.00 €

Chemical Identifiers

CAS 19646-07-2
Molecular Formula C5H4Cl2N2O
Molecular Weight (g/mol) 179.00
MDL Number MFCD02685670
InChI Key ZTHHRSBDBPCCMZ-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-methoxy-pyrimidine, 2,4-dichloro-5-methoxy pyrimidine, pyrimidine, 2,4-dichloro-5-methoxy, 2,4-dichloropyrimidin-5-yl methyl ether, 5-methoxy-2,4-dichloro pyrimidine, pubchem7076, ksc490s5l, 2,4 dichloro-5-methoxypyrimidine, 2,4-dichloro-5-methoxy-pyrimidin, 2,6-dichloro-5-methoxy pyrimidine
PubChem CID 601401
IUPAC Name 2,4-dichloro-5-methoxypyrimidine
SMILES COC1=CN=C(Cl)N=C1Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15492588
View Documents
Thermo Scientific Alfa Aesar
H56153.09
10 g
103.52 €
10g
Please to purchase this item. Need a web account? Register with us today!
15402598
View Documents
Thermo Scientific Alfa Aesar
H56153.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer’s disease.

Solubility
Slightly soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Chemical Identifiers

19646-07-2
179.00
ZTHHRSBDBPCCMZ-UHFFFAOYSA-N
601401
COC1=CN=C(Cl)N=C1Cl
C5H4Cl2N2O
MFCD02685670
2,4-dichloro-5-methoxy-pyrimidine, 2,4-dichloro-5-methoxy pyrimidine, pyrimidine, 2,4-dichloro-5-methoxy, 2,4-dichloropyrimidin-5-yl methyl ether, 5-methoxy-2,4-dichloro pyrimidine, pubchem7076, ksc490s5l, 2,4 dichloro-5-methoxypyrimidine, 2,4-dichloro-5-methoxy-pyrimidin, 2,6-dichloro-5-methoxy pyrimidine
2,4-dichloro-5-methoxypyrimidine

Specifications

19646-07-2
C5H4Cl2N2O
2,4-dichloro-5-methoxy-pyrimidine, 2,4-dichloro-5-methoxy pyrimidine, pyrimidine, 2,4-dichloro-5-methoxy, 2,4-dichloropyrimidin-5-yl methyl ether, 5-methoxy-2,4-dichloro pyrimidine, pubchem7076, ksc490s5l, 2,4 dichloro-5-methoxypyrimidine, 2,4-dichloro-5-methoxy-pyrimidin, 2,6-dichloro-5-methoxy pyrimidine
ZTHHRSBDBPCCMZ-UHFFFAOYSA-N
2,4-dichloro-5-methoxypyrimidine
601401
97%
66°C to 70°C
MFCD02685670
Slightly soluble in water.
COC1=CN=C(Cl)N=C1Cl
179.00
179
2,4-Dichloro-5-methoxypyrimidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.