Learn More
2,4-Dichloro-5-fluoropyrimidine, 97%, Thermo Scientific Chemicals
89.01 € - 456.56 €
Chemical Identifiers
CAS | 2927-71-1 |
---|---|
Molecular Formula | C4HCl2FN2 |
Molecular Weight (g/mol) | 166.96 |
MDL Number | MFCD00233551 |
InChI Key | WHPFEQUEHBULBW-UHFFFAOYSA-N |
Synonym | 2,4-dichloro-5-fluoro-pyrimidine, 5-fluoro-2,4-dichloropyrimidine, 2,6-dichloro-5-fluororacil, 2,4-dichloro-5-fluoro pyrimidine, pyrimidine, 2,4-dichloro-5-fluoro, 2,6-dichloro-5-fluoropyrimidine, 2,4-dichlor-5-fluorpyrimidin, pubchem5290, ksc204s2l, 2,4-dichloro-5fluoropyrimidine |
PubChem CID | 250705 |
IUPAC Name | 2,4-dichloro-5-fluoropyrimidine |
SMILES | FC1=CN=C(Cl)N=C1Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2927-71-1 | |
166.96 | |
WHPFEQUEHBULBW-UHFFFAOYSA-N | |
250705 | |
FC1=CN=C(Cl)N=C1Cl |
C4HCl2FN2 | |
MFCD00233551 | |
2,4-dichloro-5-fluoro-pyrimidine, 5-fluoro-2,4-dichloropyrimidine, 2,6-dichloro-5-fluororacil, 2,4-dichloro-5-fluoro pyrimidine, pyrimidine, 2,4-dichloro-5-fluoro, 2,6-dichloro-5-fluoropyrimidine, 2,4-dichlor-5-fluorpyrimidin, pubchem5290, ksc204s2l, 2,4-dichloro-5fluoropyrimidine | |
2,4-dichloro-5-fluoropyrimidine |
Specifications
2927-71-1 | |
100.0 | |
Brown to White | |
Authentic | |
Glass Bottle | |
MFCD00233551 | |
2,4-dichloro-5-fluoro-pyrimidine, 5-fluoro-2,4-dichloropyrimidine, 2,6-dichloro-5-fluororacil, 2,4-dichloro-5-fluoro pyrimidine, pyrimidine, 2,4-dichloro-5-fluoro, 2,6-dichloro-5-fluoropyrimidine, 2,4-dichlor-5-fluorpyrimidin, pubchem5290, ksc204s2l, 2,4-dichloro-5fluoropyrimidine | |
FC1=CN=C(Cl)N=C1Cl | |
166.96 | |
166.97 | |
Powder, Crystalline Powder or Low Melting Solid |
96.0 | |
38°C to 40°C | |
67°C to 68°C (6.0 Torr) | |
96% min. (GC) | |
C4HCl2FN2 | |
3261 | |
WHPFEQUEHBULBW-UHFFFAOYSA-N | |
2,4-dichloro-5-fluoropyrimidine | |
250705 | |
97% | |
2, 4-Dichloro-5-fluoropyrimidine |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- 2,4-Dichloro-5-fluoropyrimidine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.