missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-3-ethyl-6-nitrophenol, 98%, Thermo Scientific™

30.00 € - 87.00 €

Chemical Identifiers

CAS 99817-36-4
Molecular Formula C8H6Cl2NO3
Molecular Weight (g/mol) 235.04
MDL Number MFCD00270764
InChI Key YTVCECQSAPGJBB-UHFFFAOYSA-M
Synonym 2,4-dichloro-3-ethyl-6-nitrophenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, phenol, 2,4-dichloro-3-ethyl-6-nitro, 2-nitro-4,6-dichloro-5-ethylphenol, 2,4-dichloro-3-ethyl-6-nitrochloride, pubchem20442, acmc-209se5, ksc486m4b, 2,4-bis chloranyl-3-ethyl-6-nitro-phenol
PubChem CID 7020339
IUPAC Name 2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate
SMILES CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11415916
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22261.88
2.5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11425916
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B22261.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

99817-36-4
235.04
YTVCECQSAPGJBB-UHFFFAOYSA-M
7020339
CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O
C8H6Cl2NO3
MFCD00270764
2,4-dichloro-3-ethyl-6-nitrophenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, phenol, 2,4-dichloro-3-ethyl-6-nitro, 2-nitro-4,6-dichloro-5-ethylphenol, 2,4-dichloro-3-ethyl-6-nitrochloride, pubchem20442, acmc-209se5, ksc486m4b, 2,4-bis chloranyl-3-ethyl-6-nitro-phenol
2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate

Specifications

99817-36-4
C8H6Cl2NO3
UN2811
YTVCECQSAPGJBB-UHFFFAOYSA-M
2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate
7020339
98%
49°C to 51°C
MFCD00270764
2,4-dichloro-3-ethyl-6-nitrophenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, phenol, 2,4-dichloro-3-ethyl-6-nitro, 2-nitro-4,6-dichloro-5-ethylphenol, 2,4-dichloro-3-ethyl-6-nitrochloride, pubchem20442, acmc-209se5, ksc486m4b, 2,4-bis chloranyl-3-ethyl-6-nitro-phenol
CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O
235.04
236.05
2,4-Dichloro-3-ethyl-6-nitrophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c

H301-H317-H318

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 420-740-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications