missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Biphenylyl)piperazine, 95%, Thermo Scientific™

Product Code. 15473547
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105242-10-2
Molecular Formula C16H18N2
Molecular Weight (g/mol) 238.334
MDL Number MFCD05864686
InChI Key QUINYTMXFWCKTN-UHFFFAOYSA-N
Synonym 2-biphenyl-4-yl-piperazine, 2-4-phenylphenyl piperazine, 2-4-biphenylyl piperazine, 2-biphenyl-4-ylpiperazine, 2-1,1'-biphenyl-4-yl piperazine, pubchem9326, acmc-20a0oz, 2-4-biphenylyl piperazin
PubChem CID 2771747
IUPAC Name 2-(4-phenylphenyl)piperazine
SMILES C1CNC(CN1)C2=CC=C(C=C2)C3=CC=CC=C3

Specifications

Melting Point 156°C to 159°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 238.34
Percent Purity 95%
Chemical Name or Material 2-(4-Biphenylyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'