missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloro-5-methylpyrimidine, 98%, Thermo Scientific Chemicals

Product Code. 15461268
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1780-36-5
Molecular Formula C5H3Cl3N2
Molecular Weight (g/mol) 197.443
MDL Number MFCD00233525
InChI Key VTSWSQGDJQFXHB-UHFFFAOYSA-N
Synonym 5-methyl-2,4,6-trichloropyrimidine, pyrimidine, 2,4,6-trichloro-5-methyl, 2,4,6-trichlor-5-methylpyrimidin, pubchem7341, acmc-20a0tr, 2,4,6-trichloro-5, ksc179k8l, 2,6-trichloro-5-methylpyrimidine, methyl 2,4,6-trichloropyrimidine, 2,4,6-trichoro-5-methylpyrimidine
PubChem CID 232789
IUPAC Name 2,4,6-trichloro-5-methylpyrimidine
SMILES CC1=C(N=C(N=C1Cl)Cl)Cl

Specifications

Melting Point 65°C to 69°C
Quantity 250 mg
UN Number UN1759
Formula Weight 197.45
Percent Purity 98%
Chemical Name or Material 2,4,6-Trichloro-5-methylpyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'