Learn More
2,4,6,8-Tetramethylcyclotetrasiloxane, 99%, Thermo Scientific Chemicals
50.15 € - 486.42 €
Chemical Identifiers
CAS | 2370-88-9 |
---|---|
Molecular Formula | C4H12O4Si4 |
Molecular Weight (g/mol) | 236.476 |
MDL Number | MFCD00039567 |
InChI Key | WZJUBBHODHNQPW-UHFFFAOYSA-N |
Synonym | 2,4,6,8-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethylcyclotetrasiloxane, tetramethylcyclotetrasiloxane, tmcts, cyclotetrasiloxane, 2,4,6,8-tetramethyl, 2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, 1,2,5,7-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-cyclotetrasiloxan, 2,4,6,8-tetramethylcyclotetrasiloxne, 1,3,5,7-tetramethyl cyclotetrasiloxane |
PubChem CID | 6327421 |
IUPAC Name | 2,4,6,8-tetramethyl-5,7-dioxa-1,3-dioxonia-2,4-disila-6,8-disilanidacycloocta-1,3-diene |
SMILES | C[Si-]1O[Si-]([O+]=[Si]([O+]=[Si](O1)C)C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11359295
|
Thermo Scientific Alfa Aesar
L16642.06 |
5 g |
50.15 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369295
|
Thermo Scientific Alfa Aesar
L16642.14 |
25 g |
157.79 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11379295
|
Thermo Scientific Alfa Aesar
L16642.22 |
100 g |
486.42 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetramethyl-cyclotetrasiloxane is a good impregnant of photoelectric material. It is used to manufacture of modified siloxane with defined hydrogen content and chain quantity. It is also used in production of silicone polymers and acts as a precursor for the deposition of polysiloxane, cyclic siloxane, silicon dioxide, oxycarbide thin films with low dielectric constant for microelectronics and semiconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetramethyl-cyclotetrasiloxane is a good impregnant of photoelectric material. It is used to manufacture of modified siloxane with defined hydrogen content and chain quantity. It is also used in production of silicone polymers and acts as a precursor for the deposition of polysiloxane, cyclic siloxane, silicon dioxide, oxycarbide thin films with low dielectric constant for microelectronics and semiconductors.
Solubility
Miscible with most organic solvents. Immiscible with water.
Notes
Incompatible with oxidizing agents, bases and acids. Store in a cool place.
Chemical Identifiers
2370-88-9 | |
236.476 | |
WZJUBBHODHNQPW-UHFFFAOYSA-N | |
6327421 | |
C[Si-]1O[Si-]([O+]=[Si]([O+]=[Si](O1)C)C)C |
C4H12O4Si4 | |
MFCD00039567 | |
2,4,6,8-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethylcyclotetrasiloxane, tetramethylcyclotetrasiloxane, tmcts, cyclotetrasiloxane, 2,4,6,8-tetramethyl, 2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, 1,2,5,7-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-cyclotetrasiloxan, 2,4,6,8-tetramethylcyclotetrasiloxne, 1,3,5,7-tetramethyl cyclotetrasiloxane | |
2,4,6,8-tetramethyl-5,7-dioxa-1,3-dioxonia-2,4-disila-6,8-disilanidacycloocta-1,3-diene |
Specifications
134°C to 135°C | |
2370-88-9 | |
MFCD00039567 | |
1787074 | |
2,4,6,8-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethylcyclotetrasiloxane, tetramethylcyclotetrasiloxane, tmcts, cyclotetrasiloxane, 2,4,6,8-tetramethyl, 2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, 1,2,5,7-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-cyclotetrasiloxan, 2,4,6,8-tetramethylcyclotetrasiloxne, 1,3,5,7-tetramethyl cyclotetrasiloxane | |
C[Si-]1O[Si-]([O+]=[Si]([O+]=[Si](O1)C)C)C | |
236.476 | |
240.51 | |
2,4,6,8-Tetramethylcyclotetrasiloxane |
-69°C | |
C4H12O4Si4 | |
UN1993 | |
25°C (77°F) | |
WZJUBBHODHNQPW-UHFFFAOYSA-N | |
2,4,6,8-tetramethyl-5,7-dioxa-1,3-dioxonia-2,4-disila-6,8-disilanidacycloocta-1,3-diene | |
6327421 | |
99% |
Safety and Handling
GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only