missing translation for 'onlineSavingsMsg'
Learn More

2(3)-tert-Butyl-4-methoxyphenol, 96%, Thermo Scientific Chemicals

Product Code. 11485537
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2(3)-tert-Butyl-4-methoxyphenol is used as an industrial antioxidant. It is used as a food additive, preservative in food packaging, animal feed, cosmetics, rubber, and petroleum products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2(3)-tert-Butyl-4-methoxyphenol is used as an industrial antioxidant. It is used as a food additive, preservative in food packaging, animal feed, cosmetics, rubber, and petroleum products.

Solubility
Soluble in ethanol, methanol, chloroform, fats, and oils. Insoluble in water.

Notes
Store in the dark place. Store away from light, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25013-16-5
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.247
MDL Number MFCD01779059
InChI Key MRBKEAMVRSLQPH-UHFFFAOYSA-N
Synonym 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy
PubChem CID 8456
ChEBI CHEBI:76358
IUPAC Name 2-tert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=C(C=CC(=C1)OC)O

Specifications

Melting Point 55°C to 65°C
Quantity 1000 g
Sensitivity Light sensitive
Merck Index 14,1547
Solubility Information Soluble in ethanol,methanol,chloroform,fats,and oils. Insoluble in water.
Formula Weight 180.25
Percent Purity 96%
Chemical Name or Material 2(3)-tert-Butyl-4-methoxyphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.