missing translation for 'onlineSavingsMsg'
Learn More

2,3-Pyrazinedicarboxylic anhydride, 97%, Thermo Scientific Chemicals

81.00 € - 322.61 €

Chemical Identifiers

CAS 4744-50-7
Molecular Formula C6H2N2O3
Molecular Weight (g/mol) 150.09
MDL Number MFCD00179418
InChI Key AWJWCTOOIBYHON-UHFFFAOYSA-N
Synonym furo 3,4-b pyrazine-5,7-dione, 2,3-pyrazinedicarboxylic anhydride, 2,3-pyrazinecarboxylic anhydride, pyrazine-2,3-dicarboxylic anhydride, 5h,7h-furo 3,4-b pyrazine-5,7-dione, furano 3,4-b pyrazine-5,7-dione, pubchem17535, 2,3-pyrazinedicarboxylic anhydride treated bsa
PubChem CID 78482
IUPAC Name 5H,7H-furo[3,4-b]pyrazine-5,7-dione
SMILES O=C1OC(=O)C2=NC=CN=C12
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10410744
View Documents
Thermo Scientific Acros
397130050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10021334
View Documents
Thermo Scientific Acros
397130250
25 g Glass bottle
322.61 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4744-50-7
150.09
AWJWCTOOIBYHON-UHFFFAOYSA-N
78482
O=C1OC(=O)C2=NC=CN=C12
C6H2N2O3
MFCD00179418
furo 3,4-b pyrazine-5,7-dione, 2,3-pyrazinedicarboxylic anhydride, 2,3-pyrazinecarboxylic anhydride, pyrazine-2,3-dicarboxylic anhydride, 5h,7h-furo 3,4-b pyrazine-5,7-dione, furano 3,4-b pyrazine-5,7-dione, pubchem17535, 2,3-pyrazinedicarboxylic anhydride treated bsa
5H,7H-furo[3,4-b]pyrazine-5,7-dione

Specifications

4744-50-7
100.0
97%
MFCD00179418
AWJWCTOOIBYHON-UHFFFAOYSA-N
5H,7H-furo[3,4-b]pyrazine-5,7-dione
78482
97%
96.0
210°C
C6H2N2O3
furo 3,4-b pyrazine-5,7-dione, 2,3-pyrazinedicarboxylic anhydride, 2,3-pyrazinecarboxylic anhydride, pyrazine-2,3-dicarboxylic anhydride, 5h,7h-furo 3,4-b pyrazine-5,7-dione, furano 3,4-b pyrazine-5,7-dione, pubchem17535, 2,3-pyrazinedicarboxylic anhydride treated bsa
O=C1OC(=O)C2=NC=CN=C12
150.09
150.09
2, 3-Pyrazinedicarboxylic anhydride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 225-260-4

Product Identifier
  • 2,3-Pyrazinedicarboxylic anhydride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.