missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diphenyl-2-cyclopropen-1-one, 99% (dry wt.), may cont up to 5% water, Thermo Scientific Chemicals

Product Code. 11485057
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-38-4
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001311
InChI Key HCIBTBXNLVOFER-UHFFFAOYSA-N
Synonym diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec
PubChem CID 65057
ChEBI CHEBI:53074
IUPAC Name 2,3-diphenylcycloprop-2-en-1-one
SMILES O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 119°C to 121°C
Quantity 5 g
Beilstein 608049
Merck Index 14,3308
Formula Weight 206.24
Percent Purity 99%
Assay (dry wt.), may cont up to 5% water
Chemical Name or Material 2,3-Diphenyl-2-cyclopropen-1-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'