Learn More
2,3-Dimethoxybenzaldehyde, 97%, Thermo Scientific Chemicals
30.75 € - 99.09 €
Chemical Identifiers
CAS | 86-51-1 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00003309 |
InChI Key | JIVGSHFYXPRRSZ-UHFFFAOYSA-N |
Synonym | o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66 |
PubChem CID | 66581 |
IUPAC Name | 2,3-dimethoxybenzaldehyde |
SMILES | COC1=CC=CC(=C1OC)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10712685
|
Thermo Scientific Acros
115370100 |
10 g | Glass bottle |
30.75 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10705761
|
Thermo Scientific Acros
115370500 |
50 g | Glass bottle |
99.09 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
86-51-1 | |
166.18 | |
JIVGSHFYXPRRSZ-UHFFFAOYSA-N | |
66581 | |
COC1=CC=CC(=C1OC)C=O |
C9H10O3 | |
MFCD00003309 | |
o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66 | |
2,3-dimethoxybenzaldehyde |
Specifications
86-51-1 | |
100.0 | |
White to Beige | |
>110°C | |
96% min. (GC) | |
(CH3O)2C6H3CHO | |
08, I, 601 | |
JIVGSHFYXPRRSZ-UHFFFAOYSA-N | |
2,3-dimethoxybenzaldehyde | |
66581 | |
97% | |
2, 3-Dimethoxybenzaldehyde, 97% |
96.0 | |
49°C to 54°C | |
137°C (12.0 mmHg) | |
Authentic | |
C9H10O3 | |
MFCD00003309 | |
o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66 | |
COC1=CC=CC(=C1OC)C=O | |
166.18 | |
166.18 | |
Crystalline Powder, Crystals or Chunks |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-677-7
missing translation for 'tsca' : TSCA
- 2,3-Dimethoxybenzaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.