missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2',3'-Dideoxyinosine, 98%
70.10 € - 128.00 €
Chemical Identifiers
CAS | 69655-05-6 |
---|---|
Molecular Formula | C10H12N4O3 |
Molecular Weight (g/mol) | 236.23 |
MDL Number | MFCD00077728 |
InChI Key | BXZVVICBKDXVGW-JHPDDGAFNA-N |
Synonym | didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi |
PubChem CID | 50599 |
ChEBI | CHEBI:490877 |
IUPAC Name | 9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
SMILES | OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15406609
|
Thermo Scientific Chemicals
J64833.MD |
250 mg |
70.10 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15331201
|
Thermo Scientific Chemicals
J64833.ME |
500 mg |
123.00 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15496599
|
Thermo Scientific Chemicals
J64833.03 |
1 g |
128.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2?,3?-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′,3′-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.
Solubility
Soluble in DMSO or methanol
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
69655-05-6 | |
236.23 | |
BXZVVICBKDXVGW-JHPDDGAFNA-N | |
50599 | |
9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
C10H12N4O3 | |
MFCD00077728 | |
didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi | |
CHEBI:490877 | |
OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O |
Specifications
69655-05-6 | |
C10H12N4O3 | |
3619529 | |
didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi | |
BXZVVICBKDXVGW-JHPDDGAFNA-N | |
9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one | |
50599 | |
236.23 | |
Powder |
White | |
MFCD00077728 | |
143098 | |
Soluble in DMSO or methanol | |
OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O | |
236.23 | |
CHEBI:490877 | |
98% | |
2',3'-Dideoxyinosine |
Safety and Handling
missing translation for 'rtecsNumber' : NM7460700
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only