missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromobutane, (+/-) + meso, 98+%, Thermo Scientific Chemicals

Product Code. 11489552
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5408-86-6
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.916
MDL Number MFCD00000146
InChI Key BXXWFOGWXLJPPA-UHFFFAOYSA-N
Synonym butane, 2,3-dibromo, dl-2,3-dibromobutane, +/--2,3-dibromobutane, ss-butylene bromide, .+-.-2,3-dibromobutane, dibromo butane, butane,3-dibromo, beta-butylene bromide, r*,r*-+--2,3-dibromobutane, pubchem12534
PubChem CID 21508
IUPAC Name 2,3-dibromobutane
SMILES CC(C(C)Br)Br

Specifications

Density 1.76
Boiling Point 166°C to 167°C
Flash Point 123°C (253°F)
Refractive Index 1.5126
Quantity 25 g
Beilstein 1718916
Formula Weight 215.93
Percent Purity ≥98%
Chemical Name or Material 2,3-Dibromobutane, (±) + meso

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.